Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0895 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen KIAA0895
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125328
View Documents
Novus Biologicals
NBP31021325UL
25 μg
Each for $106.19
Add to cart
 
NB125327
View Documents
Novus Biologicals
NBP310213100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

KIAA0895 Polyclonal antibody specifically detects KIAA0895 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

KIAA0895
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
23366
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Mouse
hypothetical protein LOC23366, KIAA0895
The immunogen is a synthetic peptide directed towards the middle region of mouse KIAA0895 (NP_081015.1). Peptide sequence GINNLQQPWNSWIGRKKHELKPNNPTEEGLASIHSVLFRKDPFLWRAALL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit