Learn More
Abnova™ KIAA1018 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00022909T01
Catalog No. 89-949-739
Add to cart
Specifications
Specifications
293T
BC047882
pCMV-KIAA1018 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KIAA1018 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 59.6kDa
Transient overexpression cell lysate was tested with anti-KIAA1018 antibody (Mfr. No. H00022909-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human