Learn More
Aviva Systems Biology Corp KIAA1189 Antibody - middle region (ARP56188_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56188_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIAA1189. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KIAA1189. Alternate names: ermin, JN, KIAA1189ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-61753