Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1199 Polyclonal Antibody
KIAA1199 Polyclonal Antibody
Supplier: Thermo Scientific PA597125
Description
KIAA1199 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
KIAA1199 | |
Polyclonal | |
Unconjugated | |
Cemip | |
12H19.01.T7; 6330404C01Rik; 9930013L23Rik; AY007814; CCSP1; cell migration inducing hyaluronan binding protein; cell migration inducing hyaluronidase 1; cell migration inducing protein, hyaluronan binding; cell migration-inducing and hyaluronan-binding protein; CEMIP; colon cancer secreted protein 1; gb:bi851057; hyaluronan-binding protein involved in hyaluronan depolymerization; HYBID; Kiaa1199; Protein 12H19.01.T7; protein KIAA1199; RGD1305254; TMEM2L | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57214, 80982 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.33 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BI06, Q8WUJ3 | |
Cemip | |
Recombinant fusion protein containing a sequence corresponding to amino acids 180-420 of human CEMIP (NP_0012802271). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction