Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1274 purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00027143B01P
Description
Mouse polyclonal antibody raised against a full-length human KIAA1274 protein.
- High specificity
- High reactivity
Specifications
KIAA1274 purified MaxPab mouse | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human KIAA1274 protein. | |
PALD1 | |
PALD | |
Mouse | |
Affinity chromatography | |
RUO | |
27143 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_014431 | |
KIAA1274 | |
KIAA1274 (NP_055246, 1 a.a. ∼ 856 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction