Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1310 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557116
Description
Immunogen sequence: PVLFVIGQNS LQCHPEAMED FREKIRAENS LVVVGGADDN LRISKAKKKS EGLTQSMVDR CIQDEIVDFL TGVLTRAEGH MGSEPRDQDA EKKKKPR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
As part of the NSL complex it is involved in acetylation of nucleosomal histone H4 on several lysine residues and therefore may be involved in the regulation of transcription.
Specifications
KIAA1310 | |
Polyclonal | |
Unconjugated | |
KANSL3 | |
KANSL3; KAT8 regulatory NSL complex subunit 3; KIAA1310; Non-specific lethal 3 homolog; NSL complex protein NSL3; NSL3; PRTD; Rcd1; serum inhibited-related protein; SI1; Testis development protein PRTD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55683 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2N6 | |
KANSL3 | |
Recombinant protein corresponding to Human KIAA1310. Recombinant protein control fragment (Product #RP-95996). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction