Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1324 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571726
Description
KIAA1324 Polyclonal Antibody for Western Blot, IHC (P)
Expression of this gene is induced by estrogen and the encoded protein has been characterized as a transmembrane protein. The encoded protein has been found in to correlate with survival in certain carcinomas (PMID: 21102415) and may be important for cellular response to stress (PMID: 21072319). Alternative splicing results in multiple transcript variants.
Specifications
KIAA1324 | |
Polyclonal | |
Unconjugated | |
KIAA1324 | |
EIG121; Estrogen-induced gene 121 protein; KIAA1324; maba1; UNQ2426/PRO4985; UPF0577 protein KIAA1324 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
57535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q6UXG2 | |
KIAA1324 | |
KLH conjugated synthetic peptide between 704-732 amino acids from the C-terminal region of human K1324. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction