Learn More
KIAA1443 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00057594T01
Catalog No. 89-002-314
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
BC065835.1
6.8
Transient overexpression cell Lysate was tested with Anti-HOMEZ antibody (H00057594-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
HOMEZ 293T Cell Transient Overexpression Lysate(Denatured)
58.7
pCMV-HOMEZ full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.