Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1530 Mouse anti-Human, Clone: GT816, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527844

Catalog No. PIMA527844


Add to cart

Description

Description

Positive Control: HeLa, A375 Predicted Reactivity: Mouse (83%), Rat (83%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by KIAA1530 appears to be involved in ubiquitination and dephosphorylation of RNA polymerase II subunits that stall after UV irradiation. The encoded protein interacts with several members of the nucleotide excision repair complex, and is thought to be involved in the transcription-coupled nucleotide excision repair (TC-NER) pathway to help remove lesions in the DNA that block transcription. Defects in KIAA1530 can cause UV-sensitive syndrome 3. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

KIAA1530
Monoclonal
1 mg/ml
PBS with no preservative
Q2YD98
UVSS3, UV-stimulated scaffold protein A, KIAA1530
Mouse
IgG1
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
57654
Western Blot
GT816
Unconjugated
UVSSA
Liquid
UVSSA
Recombit protein encompassing a sequence within the N-terminus region of human UVSSA. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit