Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1549 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569501
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
KIAA1549 is a protein coding gene.
Specifications
KIAA1549 | |
Polyclonal | |
Unconjugated | |
KIAA1549 | |
D630045J12Rik; KIAA1549; KIAA1549 ortholog; mKIAA1549; RIKEN cDNA D630045J12 gene; UPF0606 protein KIAA1549; UPF0606 protein KIAA1549 homolog; UPF0606 protein KIAA1549; UPF0606 protein KIAA1549 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
330286, 57670 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q68FD9, Q9HCM3 | |
D630045J12Rik, KIAA1549 | |
synthetic peptide directed towards the middle region of human KIAA1549 (aa 1882-1931). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction