Learn More
Abnova™ KIAA1604 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00057703T02
Catalog No. 89-949-817
Add to cart
Description
Description
Specifications
Specifications
293T
BC093952
pCMV-KIAA1604 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KIAA1604 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 105.5kDa
Transient overexpression cell lysate was tested with anti-KIAA1604 antibody (Mfr. No. H00057703-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human