Learn More
Abnova™ KIAA1627 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00057721T01
Catalog No. 89-949-822
Add to cart
Specifications
Specifications
Western Blot
293T
NM_020961
pCMV-METTL14 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
METTL14 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 52.2kDa
Transient overexpression cell lysate was tested with anti-KIAA1627 antibody (Mfr. No. H00057721-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)