Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA2013 Polyclonal Antibody
KIAA2013 Polyclonal Antibody
Supplier: Thermo Scientific PA562664
Description
KIAA2013 Polyclonal Antibody for Western Blot, IHC

Specifications
KIAA2013 | |
Polyclonal | |
Unconjugated | |
KIAA2013 | |
KIAA2013; RP5-1077B9.1; uncharacterized protein KIAA2013 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYS2 | |
KIAA2013 | |
Recombinant protein corresponding to Human KIAA2013. Recombinant protein control fragment (Product #RP-100869). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction