Learn More
Kidde Fire Fighting Premier Lite Fire Hoses
Manufacturer:  NATIONAL FOAM ANGUS FIRE 011067
 View more versions of this product
Catalog No. 19-805-342
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
8.96cm
3 in.
48.26cm
Nylon
24 lb.
Double jacket fire hose with Urethane lining
Yellow
19 in.
7.62cm
800psi
10.88kg