Learn More
Kidde™ Fire Trainer Type-II Smoke Fluid
Type II smoke fluid used in Kidde smoke generators
$248.00 - $1453.00
Specifications
Product Type Type-II smoke fluid
For Use With (Application) Producing a long lasting, low moisture fog which is perfect for high heat training environments
For Use With (Equipment) Smoke generator
View More Specs
Products 3
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
19-139-651
View Documents
Kidde™
1A1002017001
1.5 gal.
Each for $248.00
Add to cart
19-139-652
View Documents
Kidde™
1A002017002
5 gal.
Each for $304.50
Add to cart
19-139-653
View Documents
Kidde™
1A1002017003
55 gal.
Each for $1,453.00
Add to cart
Description
Description
Made from the highest quality pharmaceutical grade chemicals and perfect for high heat training environments.
  • Mineral oil type
  • Non-toxic, colorless, odorless, non-flammable
  • Produces long lasting, low moisture fog perfect for high heat training environments
Specifications
Specifications
Type-II smoke fluid
Producing a long lasting, low moisture fog which is perfect for high heat training environments
Smoke generator