Kidde™ Fire Trainer Type-II Smoke Fluid Available on GSA/VA Contract for Federal Government customers only.
Type II smoke fluid used in Kidde smoke generators
Manufacturer: KIDDE FIRE TRAINERS LLC 1A002017002
 View more versions of this product
Catalog No. 19-139-652
  Check Availability
Add to Cart
    Specifications
    Capacity 5 gal.
    For Use With (Application) Producing a long lasting, low moisture fog which is perfect for high heat training environments
    For Use With (Equipment) Smoke generator
    Product Type Type-II smoke fluid
  • Description & Specifications
Related Products
Product Content Feedback