Learn More
Kidde™ Fire Trainer Type-II Smoke Fluid Available on GSA/VA Contract for Federal Government customers only.
Type II smoke fluid used in Kidde smoke generators
Manufacturer:  KIDDE FIRE TRAINERS LLC 1A1002017003
 View more versions of this product
Catalog No. 19-139-653
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
55 gal.
Producing a long lasting, low moisture fog which is perfect for high heat training environments
Smoke generator
Type-II smoke fluid