Learn More
Kidde™ Mobile Aircraft FIRETRAINER™ MA-500
Reliable, convenient and economical mobile aircraft training
$683.50 - $1554400.00
Specifications
For Use With (Equipment) Mobile aircraft Firetrainer™ MA-500
View More Specs
Products 6
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
19-139-667
View Documents
Kidde™
MASS
Full Stainless Steel option
Each for $111,350.00
Add to cart
19-139-668
View Documents
Kidde™
MA500FMZ1
Full maintenance
Each for $37,050.00
Add to cart
19-038-684
View Documents
Kidde™
MA500MAFTD
US navy configuration
Each for $1,554,400.00
Add to cart
19-139-434
View Documents
Kidde™
MASPC
Shore power connection
Each for $683.50
Add to cart
19-139-435
View Documents
Kidde™
T500DATA1
Data storage and retrieval package
Each for $22,780.00
Add to cart
19-139-450
View Documents
Kidde™
MAWLESS
Wireless control
Each for $5,470.00
Add to cart
Description
Description
Live Fire Training System™ simulates fire emergencies on military fighter and transport aircrafts and is currently in use around the world, providing user-friendly training to hundreds of aircraft rescue firefighters.
  • Road-mobile aircraft fire simulator designed to expose trainees to the situations encountered in aircraft rescue fire fighting (ARFF)
  • Utilizes environmentally friendly propane as its fuel source
  • Equipped with onboard control and safety systems
  • Fully configurable to meet numerous training curricula, including internal and external fire scenarios, forced entry panels, internal seat/cargo configurations, typical and emergency entryways, and wing and tail engines and configurations
Order Info
Accessories, options and warranties available separately. FireTrainer available for use in Zone 1 (within 48 contiguous states). The Kidde Mobile Aircraft FIRETRAINER MA-500 is also available for purchase through Fisher Safety's GSA Advantage!™ Schedule 84.
Specifications
Specifications
Mobile aircraft Firetrainer™ MA-500