Learn More
Kidde™ Mobile HazMat Trainer
Mobile HazMat Trainer fully self-sufficient hazardous materials training device, designed to expose trainees to situations encountered by HazMat response personnel
Manufacturer: Kidde™ S 10 CH
Description
Utilizes environmentally friendly propane for live fires and air and water to simulate various leaks and spills
- On-board propane supply and storage system
- Composed of air compressor and air/water distribution system
- Fully configurable to meet any number of training curricula, and can be customized to meet specific needs
- Portability allows for training at numerous locations without cost of repetitive infrastructure
Specifications
Chlorine 1 Ton training end | |
Mobile Hazmat trainer |