Learn More
Kidde™ Mobile Structural FIRETRAINER™ T-4000
Permits live-fire training in a safe, controlled and environmentally sound manner
$726.50 - $493850.00
Specifications
Certifications/Compliance NFPA 1402
For Use With (Equipment) Mobile structural Firetrainer™ T-4000
Includes 45 ft. Container based, mobile fire training trailer, on site start up and operation/maintenance training, operation and maintenance manuals
View More Specs
Includes: 45 ft. container-based, mobile fire training trailer, On-site start-up and operation/maintenance training, Operation and maintenance manuals.
Products 19
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
19-139-541
View Documents
Kidde™
T4000CONSP
Removable L-shaped confined space tube
Each for $12,680.00
Add to cart
19-139-540
View Documents
Kidde™
T40000CFTOWER
Modular confined space tower
Each for $252,450.00
Add to cart
19-038-683
View Documents
Kidde™
T4000DR
Movable door partition
Each for $1,292.00
Add to cart
19-038-682
View Documents
Kidde™
T4000WL
Movable wall partition
Each for $1,175.00
Add to cart
19-139-801
View Documents
Kidde™
T4000453 Z1
Fire training system
Each for $493,850.00
Add to cart
19-139-802
View Documents
Kidde™
T4000R Z1
Flareup/rollover option
Each for $13,800.00
Add to cart
19-139-803
View Documents
Kidde™
T40002FLFIREZ1
Fire effect in second story
Each for $33,050.00
Add to cart
19-139-806
View Documents
Kidde™
T4000MOCKZ1
Interchangeable mockup
Each for $2,620.00
Add to cart
19-139-809
View Documents
Kidde™
T4000SPC Z1
Shore power connection
Each for $726.50
Add to cart
19-139-805
View Documents
Kidde™
T4000WLESSZ1
Wireless control
Each for $5,810.00
Add to cart
19-139-810
View Documents
Kidde™
T4000SPARESZ1
Spare repair part set
Each for $1,816.00
Add to cart
19-139-808
View Documents
Kidde™
T4000ELPZ1
Exterior lighting
Each for $3,510.00
Add to cart
19-139-804
View Documents
Kidde™
T4000SSZ1
Sound system
Each for $16,590.00
Add to cart
19-139-807
View Documents
Kidde™
T4000SPZ1
Standpipe and sprinkler system
Each for $3,880.00
Add to cart
19-139-811
View Documents
Kidde™
T4000GRA Z1
Graphics package option
Each for $3,760.00
Add to cart
19-139-812
View Documents
Kidde™
T4000FM Z1
Full Maintenance Warranty
Each for $16,430.00
Add to cart
19-139-813
View Documents
Kidde™
T4000PM Z1
Preventive Maintenance Warranty
Each for $9,620.00
Add to cart
19-139-429
View Documents
Kidde™
T4000 45 3
Fire trainer with 3 fires
Each for $493,850.00
Add to cart
19-139-449
View Documents
Kidde™
T4000 FEWIN
Forcible entry window prop
Each for $911.50
Add to cart
Description
Description
This turnkey training system offers a convenient means to train students locally in basic NFPA 1001 firefighting skills and advanced firefighting drills, allowing for more frequent training and increased emergency preparedness. The T-4000 utilizes a heavy-duty, corrosion-resistant 45 ft. ISO container as its base structure and 53 ft. trailer chassis.
One, two, or three propane-fueled, programmable AquaMesh™ fires with automatic agent detection features:
  • Two interchangeable multi-prop mockups
  • Flashover-rollover effect
  • Interior movable walls and door
  • Pitched roof ventilation prop
  • Smoke generation system
  • Ship's ladder with rooftop hatch
  • 200 gal. on-board propane storage
  • On-board 12kW generator
  • 10-gauge smooth exterior cladding
  • Diamond plate floor and roof
Order Info
Accessories, options and warranties available separately. FireTrainer available for use in Zone 1 (within 48 contiguous states). 24/7 technical phone support available. The Kidde Mobile Structural FIRETRAINER T-4000 is also available for purchase through Fisher Safety's GSA Advantage!™ Schedule 84.
Compliance
NRTL certification per NFPA 1402. Third-party safety analysis report.
Warranty and Services
One year on parts
Specifications
Specifications
NFPA 1402
Mobile structural Firetrainer™ T-4000
45 ft. Container based, mobile fire training trailer, on site start up and operation/maintenance training, operation and maintenance manuals