Kidde™ Mobile Structural FIRETRAINER™ T-4000 Available on GSA/VA Contract for Federal Government customers only.
Permits live-fire training in a safe, controlled and environmentally sound manner
Manufacturer: KIDDE FIRE TRAINERS LLC T4000WL
 View more versions of this product
Catalog No. 19-038-682
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1402
    For Use With (Equipment) Mobile structural Firetrainer™ T-4000
    Includes 45 ft. Container based, mobile fire training trailer, on site start up and operation/maintenance training, operation and maintenance manuals
    Product Type Movable wall partition
  • Description & Specifications
Related Products
Product Content Feedback