Learn More
Kidde™ Mobile Structural FIRETRAINER™ T-4000 Available on GSA/VA Contract for Federal Government customers only.
Permits live-fire training in a safe, controlled and environmentally sound manner
Manufacturer:  KIDDE FIRE TRAINERS LLC T4000R Z1
 View more versions of this product
Catalog No. 19-139-802
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1402
Mobile structural Firetrainer™ T-4000
45 ft. Container based, mobile fire training trailer, on site start up and operation/maintenance training, operation and maintenance manuals
Flareup/rollover option