Learn More
Kidde™ Portable Hose Line FIRETRAINER™ O-100
Reliable, convenient and economical outdoor fire training
$1709.00 - $289300.00
Specifications
Certifications/Compliance | NFPA 1001 |
---|---|
For Use With (Equipment) | Portable hoseline Firetrainer™ |
Includes: 50 ft. AC power cable.
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-139-649
|
Kidde™
EXPLGENZ1 |
Explosion noise generator |
Each for $7,260.00
|
|
19-139-615
|
Kidde™
O100CTR |
Self-contained transport trailer |
Each for $54,950.00
|
|
19-139-647
|
Kidde™
O1003 TANKER Z1 |
Mobile Hazmat tanker |
Each for $289,300.00
|
|
19-139-629
|
Kidde™
O100CSTND |
Control stand |
Each for $13,800.00
|
|
19-139-618
|
Kidde™
O100BBQ |
BBQ grill |
Each for $13,080.00
|
|
19-139-625
|
Kidde™
O100EMP |
Motor generator |
Each for $5,810.00
|
|
19-139-631
|
Kidde™
O100HELO |
Helicopter mockup |
Each for $67,450.00
|
|
19-139-624
|
Kidde™
O100TRNS |
Electrical transformer |
Each for $5,810.00
|
|
19-139-630
|
Kidde™
O100WLESS |
Wireless control for 0-100 |
Each for $5,810.00
|
|
19-139-623
|
Kidde™
O100SFL |
Split flange pipe |
Each for $8,360.00
|
|
19-139-617
|
Kidde™
O100XMT |
Christmas tree (industrial pipe) |
Each for $5,450.00
|
|
19-139-650
|
Kidde™
O100WLESSZ1 |
Wireless control for O-100 Tanker |
Each for $5,810.00
|
|
19-139-614
|
Kidde™
O100PLUS |
Hose Line Trainer O-100 Plus; 112-square-foot burn area |
Each for $60,800.00
|
|
19-139-613
|
Kidde™
O100 |
Hose Line Trainer O-100; 24-square foot burn area |
Each for $23,850.00
|
|
19-139-616
|
Kidde™
O100CAR |
Car |
Each for $36,600.00
|
|
19-139-619
|
Kidde™
O100PCP |
Propane cylinder |
Each for $5,450.00
|
|
19-139-626
|
Kidde™
O100FBK |
Flame bar kit |
Each for $3,390.00
|
|
19-139-627
|
Kidde™
O100PBONLY |
Pilot box |
Each for $17,430.00
|
|
19-139-621
|
Kidde™
O100PV |
Pressure Vessel prop; 250 gal. |
Each for $8,360.00
|
|
19-139-620
|
Kidde™
O100DUMP |
Commercial dumpster |
Each for $19,010.00
|
|
19-139-628
|
Kidde™
O100MULTI |
Multiple control kit |
Each for $13,800.00
|
|
19-139-414
|
Kidde™
O100 HEXT |
Hoseline extension kit |
Each for $1,709.00
|
|
19-139-416
|
Kidde™
O100 F16 |
F16 mockup |
Each for $82,250.00
|
|
19-139-415
|
Kidde™
O100 PANONLY |
Hoseline trainer pan |
Each for $6,040.00
|
|
19-139-424
|
Kidde™
0100 DSFL |
Dual split pipe flange |
Each for $11,170.00
|
|
19-139-437
|
Kidde™
O100 PV500 |
Pressure Vessel prop; 500 gal. |
Each for $14,240.00
|
|
19-139-438
|
Kidde™
O100 LPG |
Propane supply trailer |
Each for $25,900.00
|
|
19-139-444
|
Kidde™
O100 TRUCK |
Portable truck |
Each for $36,600.00
|
|
Description
The Hose Line FireTrainer O-100 is a convenient means of training students in the basic firefighting skills. The trainer can be used as a standalone burn pan to replicate fuel spill or ground fires in addition to several other types of fire scenarios. It provides hot, challenging fires to train both new recruits and seasoned firefighters.
- Can be used as a standalone training tool to replicate fuel spill or ground fires in addition to several other types of fire scenarios
- Independent constant pilot flame guarantees each fire ignites on command
- Main flame control
- Hand-held operator/instructor “deadman” button on the control instantly stops fire and ensures safety during training exercises
- Completely portable unit that can be setup and operated by one person
- Fueled by clean burning propane
- Stainless-steel construction
- Low set-up and clean-up, more training time
- Weight: approximately 200 lbs. Power: 110VAC. Fire output: 65,000,000 btu/hr (20 MW)
Order Info
Optional props, accessories and warranties are available (sold separately).
Compliance
FireTrainer O-100 Plus meets NFPA 1001 training requirements.
Warranty and Services
One year on parts.
Specifications
NFPA 1001 | |
Portable hoseline Firetrainer™ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.