Learn More
Kidde™ Structural FIRETRAINER™ T-500
Essential, operator-controlled live fire training
$3870.00 - $200950.00
Specifications
For Use With (Equipment) Structural Firetrainer™ T-500
Includes Installation, on-site start-up service and testing by a factory technician and operation and maintenance training, equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensor, temperature sensor, smoke cabinet and operator pendant
View More Specs
Includes: Includes installation, on-site start-up service and testing by a factory technician, and operation and maintenance training. Equipment: water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensors, temperature sensor, smoke cabinet and operator pendant.
Requires: Natural gas or propane fuel supply and 208V/3PH/60Hz-30 A or 415V/3PH/50Hz-20 A power supply.
Products 14
Catalog Number Mfr. No. Certifications/Compliance Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Certifications/Compliance Product Type Price Quantity & Availability  
19-139-600
View Documents
Kidde™
T500FP1Z1
NFPA 54 CGA 149.3 Structural Firetrainer™ with one fire in one room
Each for $200,950.00
Add to cart
19-139-603
View Documents
Kidde™
T500FZ1
NFPA 54 CGA 149.3 Structural Firetrainer™ with flashover effect
Each for $53,300.00
Add to cart
19-139-601
View Documents
Kidde™
T500FP2Z1
NFPA 54 CGA 149.3 Structural Firetrainer™ with second fire in a different room
Each for $161,000.00
Add to cart
19-139-605
View Documents
Kidde™
T500IP6Z1
NFPA 54 CGA 149.3 Six Interchangeable Props
Each for $20,580.00
Add to cart
19-139-602
View Documents
Kidde™
T500MAJZ1
NFPA 54 CGA 149.3 Structural Firetrainer™ with second major fire in one room
Each for $127,100.00
Add to cart
19-139-607
View Documents
Kidde™
T500VZ1
NFPA 54 CGA 149.3 Structural Firetrainer™ with vent system
Each for $41,200.00
Add to cart
19-139-606
View Documents
Kidde™
T500WLESSZ1
NFPA 54 CGA 149.3 Wireless control
Each for $5,810.00
Add to cart
19-139-604
View Documents
Kidde™
T500RZ1
NFPA 54 CGA 149.3 Structural Firetrainer™ with flare-up/rollover effect
Each for $16,950.00
Add to cart
19-139-405
View Documents
Kidde™
T500FP1
NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402 Structural Firetrainer™ first fire with smoke
Each for $200,950.00
Add to cart
19-139-411
View Documents
Kidde™
T500ATT
NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402 Structural Firetrainer™ with attic fire standalone
Each for $148,050.00
Add to cart
19-139-410
View Documents
Kidde™
T500DUCT
NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402 Structural Firetrainer™ with hidden attic/duct fire
Each for $148,050.00
Add to cart
19-139-409
View Documents
Kidde™
T500FP2
NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402 Structural Firetrainer™ additional fire with smoke
Each for $161,000.00
Add to cart
19-139-442
View Documents
Kidde™
T500FM2
NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402 Structural Firetrainer™ with Full maintenance
Each for $3,870.00
Add to cart
19-139-448
View Documents
Kidde™
T500V
NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402 Standalone vent system
Each for $41,200.00
Add to cart
Description
Description
The Structural T-500 provides essential gas-fueled structural fire training.
  • Instant ignition, shutdown and re-ignition of fires using a handheld control pendant
  • Large, realistic hot fires under direct operator control
  • Interchangeable fire mockups for versatility
  • Powerful smoke generator with every fire can be used in conjunction or independent of the fires for search and rescue training and breathing apparatus training
  • Panel-mounted system controls
  • Handheld control pendant with intuitive lighted feedback
  • PLC computer monitoring of safety conditions and systems
  • Self-diagnostics for easy use
  • Environmentally sound clean-burning, gas-fueled fires are environmentally sound
Order Info
Accessories and options available separately.
Compliance
NRTL certification per NFPA. Valve cabinet meets NFPA 54, CGA 149.3 standards; pilot burner meets FM and CSA standards; electrical control cabinet meets NFPA 70 standards; flammable gas and temperature sensors meet NFPA 1402 standards.
Warranty and Services
One year on parts with 24/7 toll-free support.
Specifications
Specifications
Structural Firetrainer™ T-500
Installation, on-site start-up service and testing by a factory technician and operation and maintenance training, equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensor, temperature sensor, smoke cabinet and operator pendant