Kidde™ Structural FIRETRAINER™ T-500 Available on GSA/VA Contract for Federal Government customers only.
Essential, operator-controlled live fire training
Manufacturer: KIDDE FIRE TRAINERS LLC T500FP1
 View more versions of this product
Catalog No. 19-139-405
  Check Availability
Add to Cart
Required Accessories : Natural gas or propane fuel supply and 208V/3PH/60Hz-30 A or 415V/3PH/50Hz-20 A power supply.
    Specifications
    Certifications/Compliance NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402
    Includes Installation, on-site start-up service and testing by a factory technician and operation and maintenance training. Equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensors, temperature sensor, smoke
    Product Type Structural Firetrainer™ first fire with smoke
    For Use With (Equipment) Structural Firetrainer™ T-500
  • Description & Specifications
Related Products
Product Content Feedback