Kidde™ Structural FIRETRAINER™ T-500 Available on GSA/VA Contract for Federal Government customers only.
Essential, operator-controlled live fire training
Manufacturer: KIDDE FIRE TRAINERS LLC T500FP2
 View more versions of this product
Catalog No. 19-139-409
  Check Availability
Add to Cart
Required Accessories : Natural gas or propane fuel supply and 208V/3PH/60Hz-30 A or 415V/3PH/50Hz-20 A power supply.
    Specifications
    Certifications/Compliance NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402
    Includes Installation, on-site start-up service and testing by a factory technician and operation and maintenance training. Equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensors, temperature sensor, smoke
    For Use With (Equipment) Structural Firetrainer™ T-500
    Product Type Structural Firetrainer™ additional fire with smoke
  • Description & Specifications
Related Products
Product Content Feedback