Learn More
Kidde™ Structural FIRETRAINER™ T-500 Available on GSA/VA Contract for Federal Government customers only.
Essential, operator-controlled live fire training
Manufacturer:  KIDDE FIRE TRAINERS LLC T500ATT
 View more versions of this product
Catalog No. 19-139-411
  Check Availability
Add to cart
Requires : Natural gas or propane fuel supply and 208V/3PH/60Hz-30 A or 415V/3PH/50Hz-20 A power supply.
Specifications Description & Specifications
Specifications
NFPA 54, CGA 149.3, FM, CSA, NFPA 70, NFPA 1402
Structural Firetrainer™ T-500
Installation, on-site start-up service and testing by a factory technician and operation and maintenance training. Equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensors, temperature sensor, smoke
Structural Firetrainer™ with attic fire standalone