Learn More
Kidde™ Structural FIRETRAINER™ T-500 Available on GSA/VA Contract for Federal Government customers only.
Essential, operator-controlled live fire training
Manufacturer:  KIDDE FIRE TRAINERS LLC T500FP1Z1
 View more versions of this product
Catalog No. 19-139-600
  Check Availability
Add to cart
Requires : Natural gas or propane fuel supply and 208V/3PH/60Hz-30 A or 415V/3PH/50Hz-20 A power supply.
Specifications Description & Specifications
Specifications
NFPA 54 CGA 149.3
Structural Firetrainer™ T-500
Installation, on-site start-up service and testing by a factory technician and operation and maintenance training, equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensor, temperature sensor, smoke cabinet and operator pendant
Structural Firetrainer™ with one fire in one room