Kidde™ Structural FIRETRAINER™ T-500 Available on GSA/VA Contract for Federal Government customers only.
Essential, operator-controlled live fire training
Manufacturer: KIDDE FIRE TRAINERS LLC T500IP6Z1
 View more versions of this product
Catalog No. 19-139-605
  Check Availability
Add to Cart
Required Accessories : Natural gas or propane fuel supply and 208V/3PH/60Hz-30 A or 415V/3PH/50Hz-20 A power supply.
    Specifications
    Certifications/Compliance NFPA 54 CGA 149.3
    For Use With (Equipment) Structural Firetrainer™ T-500
    Includes Installation, on-site start-up service and testing by a factory technician and operation and maintenance training, equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensor, temperature sensor, smoke cabinet and operator pendant
    Product Type Six Interchangeable Props
  • Description & Specifications
Related Products
Product Content Feedback