Learn More
Kidde™ Structural FIRETRAINER™ T-500 Available on GSA/VA Contract for Federal Government customers only.
Essential, operator-controlled live fire training
Manufacturer:  KIDDE FIRE TRAINERS LLC T500VZ1
 View more versions of this product
Catalog No. 19-139-607
  Check Availability
Add to cart
Requires : Natural gas or propane fuel supply and 208V/3PH/60Hz-30 A or 415V/3PH/50Hz-20 A power supply.
Specifications Description & Specifications
Specifications
Structural Firetrainer™ with vent system
NFPA 54 CGA 149.3
Structural Firetrainer™ T-500
Installation, on-site start-up service and testing by a factory technician and operation and maintenance training, equipment Water bath burn platform, valve cabinet, pilot burner, electrical control cabinet, flammable gas sensor, temperature sensor, smoke cabinet and operator pendant