Learn More
KIF11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA132455
Catalog No. PIPA132455
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-32455 detects Eg5 from human samples. PA1-32455 has been successfully used in immunofluorescence, immunoprecipitation, Western blot applications. The PA1-32455 immunogen is: Produced by immunization of rabbits with a recombit segment of the coiled-coil domain of the human protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KIF11
Polyclonal
KIF11
EG5, HKSP, KNSL1, TRIP5
Rabbit
100 μL
Primary
Human, Mammalia
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
P52732, Q6P9P6
KIF11
Produced by immunization of rabbits with a recombit segment of the coiled-coil domain of the human protein.
RUO
16551, 171304, 3832
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations
Recommendations