Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF16B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30452320UL
Description
KIF16B Polyclonal specifically detects KIF16B in Human samples. It is validated for Western Blot.Specifications
KIF16B | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
C20orf23, chromosome 20 open reading frame 23, dJ971B4.1, FLJ20135, KIAA1590, kinesin family member 16B, kinesin-like protein KIF16B, KISC20ORF, SNX23kinesin motor protein, sorting nexin 23, Sorting nexin-23 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human KIF16B (NP_078980.3). | |
20 μL | |
Signal Transduction | |
55614 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction