Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF18A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen KIF18A
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096306
View Documents
Novus Biologicals
NBP305120100UL
100 μg Each for $320.71
Add to cart
 
NB096307
View Documents
Novus Biologicals
NBP30512020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

KIF18A Polyclonal antibody specifically detects KIF18A in Human samples. It is validated for Western Blot
Specifications

Specifications

KIF18A
Unconjugated
Rabbit
Human
DKFZP434G2226, kinesin family member 18A, kinesin-like protein KIF18A, Marrow stromal KIF18A, MS-KIF18A
Recombinant fusion protein containing a sequence corresponding to amino acids 599-898 of human KIF18A (NP_112494.3).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
81930
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit