Learn More
Abnova™ KIF19 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00124602T01
Catalog No. 89-949-870
Add to cart
Specifications
Specifications
293T
BC093631
pCMV-KIF19 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KIF19 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 62kDa
Transient overexpression cell lysate was tested with anti-KIF19 antibody (Mfr. No. H00124602-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human