Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen KIF1B
Concentration 0.36 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-0101
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
152631AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

The kinesins constitute a large family of microtubule-dependent motor proteins, which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events including endocytosis and transcytosis. KIF1B is a member of the KIF1/Unc104 family of kinesin-like proteins that are involved in the transport of mitochondria or synaptic vesicles in axons. KIF1B is an amino-terminal-type motor protein that is ubiquitously expressed, with the most abundant levels in differentiated nerve cells. The human KIF1B gene maps to chromosome 1p36.2. Defects in axonal transport due to mutations at the KIF1B gene can underlie the human peripheral neuropathy phenotype. The mouse KIF1B gene generates an alternatively spliced transcript, which produces two isoforms.
Specifications

Specifications

KIF1B
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
IgG
-20°C
0.36 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit