Learn More
KIF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152631AP
Catalog No. 50-173-0101
Add to cart
Description
Description
The kinesins constitute a large family of microtubule-dependent motor proteins, which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events including endocytosis and transcytosis. KIF1B is a member of the KIF1/Unc104 family of kinesin-like proteins that are involved in the transport of mitochondria or synaptic vesicles in axons. KIF1B is an amino-terminal-type motor protein that is ubiquitously expressed, with the most abundant levels in differentiated nerve cells. The human KIF1B gene maps to chromosome 1p36.2. Defects in axonal transport due to mutations at the KIF1B gene can underlie the human peripheral neuropathy phenotype. The mouse KIF1B gene generates an alternatively spliced transcript, which produces two isoforms.
Specifications
Specifications
KIF1B
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60333, O88658, Q60575
CMT2, CMT2A, CMT2A1, HMSNII, KIAA0591, KIAA1448, KIF1B, kinesin family member 1B, Kinesin like protein KIF1B, KLP
Rabbit
IgG
150 μL
-20°C
Primary
117548, 16561, 23095
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
KIF1B
Liquid
Kif1b
KIF1B Fusion Protein Ag7263
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat