Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF21A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540809
Description
Peptide sequence: KEKRKKKSVA GKEDNTDTDQ EKKEEKGVSE RENNELEVEE SQEVSDHEDE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 86%; Yeast: 77%.
KIF21A gene ontology annotations related to this gene include microtubule-based movement.
Specifications
KIF21A | |
Polyclonal | |
Unconjugated | |
Kif21a | |
CFEOM1; FEOM1; FEOM3A; FLJ20052; KIAA1708; KIF2; Kif21a; kinesin family member 21A; kinesin-like protein KIF2; Kinesin-like protein KIF21A; N-5 kinesin; putative LOC540076 protein; renal carcinoma antigen NY-REN-62; Unknown (protein for IMAGE:8217042) | |
Rabbit | |
Affinity chromatography | |
RUO | |
55605 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z4S6 | |
Kif21a | |
Synthetic peptide directed towards the N-terminal of human KIF21A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction