Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF25 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550403
Description
The antibody detects endogenous levels of total KIF25 protein.
KIF25 Polyclonal antibody specifically detects KIF25 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
KIF25 | |
Polyclonal | |
Unconjugated | |
KIF25 | |
KIF25; kinesin family member 25; kinesin-like 3; kinesin-like protein 3; Kinesin-like protein KIF25; KNSL3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3834 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UIL4 | |
KIF25 | |
Fusion protein corresponding to residues near the N terminal of human kinesin family member 25 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction