Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF25 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18003520UL

 View more versions of this product

Catalog No. NBP18003520


Add to cart

Description

Description

KIF25 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

KIF25
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
KIF25
Synthetic peptide directed towards the N terminal of human KIF25. Peptide sequence: TWTSGQLQREKQARPGSGAVLAFPDDKDLRVYGPAESQSAVFGDVCPLLT
Immunogen affinity purified
RUO
Primary
3834
Western Blot
Western Blot 1:1000
NP_005346
kinesin family member 25
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit