Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555186
Description
Immunogen sequence: LPESPCLSPL KPLKPHFGDI QEGIYVAIQR SDKRIHLAVV TEINRENYWV TVEWVEKAVK KGKKIDLETI LLLNPALDSA EHP Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
KIF2B is the plus end-directed microtubule-dependent motor required for spindle assembly and chromosome movement. KIF2B has microtubule depolymerization activity.
Specifications
KIF2B | |
Polyclonal | |
Unconjugated | |
KIF2B | |
1700063D03Rik; EMBL:AAH83841.1, ECO:0000312; KIF2B; kif2b {ECO:0000312; kinesin family member 2B; kinesin protein; kinesin-like protein KIF2B; LOW QUALITY PROTEIN: kinesin-like protein KIF2B; RGD:1359248}; testis tissue sperm-binding protein Li 82P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N4N8 | |
KIF2B | |
Recombinant protein corresponding to Human KIF2B. Recombinant protein control fragment (Product #RP-93276). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction