Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen KIF2B
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094608
View Documents
Novus Biologicals
NBP305100100UL
100 μg
Each for $320.71
Add to cart
 
NB094609
View Documents
Novus Biologicals
NBP30510020UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

KIF2B Polyclonal antibody specifically detects KIF2B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

KIF2B
Unconjugated
Rabbit
Human, Mouse, Rat
FLJ53902, kinesin family member 2B, kinesin protein, kinesin-like protein KIF2B
Recombinant fusion protein containing a sequence corresponding to amino acids 544-673 of human KIF2B (NP_115948.4).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
84643
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit