Learn More
Abnova™ KIF2C 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00011004T02
Catalog No. 89-949-888
Add to cart
Description
Description
Specifications
Specifications
293T
BC014924
pCMV-KIF2C full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KIF2C 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 81.3kDa
Transient overexpression cell lysate was tested with anti-KIF2C antibody (Mfr. No. H00011004-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human