Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF2C Mouse anti-Human, Mouse, Rat, Monkey, PE/Atto594, Clone: OTI1C5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271667PEATT594

Catalog No. N27167PE594


Add to cart

Description

Description

KIF2C Monoclonal antibody specifically detects KIF2C in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

KIF2C
OTI1C5
Flow Cytometry
Affinity Purified, Conjugated
KIF2C
Full length recombinant protein of human KIF2C (NP_006836) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Cell Biology, Cytoskeleton Markers, DNA Repair
Primary
11004.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit