Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen KIF3B
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094624
View Documents
Novus Biologicals
NBP305143100UL
100 μg
Each for $320.71
Add to cart
 
NB094625
View Documents
Novus Biologicals
NBP30514320UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

KIF3B Polyclonal antibody specifically detects KIF3B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

KIF3B
Unconjugated
Rabbit
Human, Mouse, Rat
HH0048, KIAA0359Microtubule plus end-directed kinesin motor 3B, kinesin family member 3B, kinesin-like protein KIF3B
Recombinant fusion protein containing a sequence corresponding to amino acids 663-747 of human KIF3B (NP_004789.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
9371
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit