Learn More
Abnova™ KIF4A 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00024137T01
Catalog No. 89-949-899
Add to cart
Specifications
Specifications
Western Blot
293T
BC050548.1
pCMV-KIF4A full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
KIF4A 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 124.08kDa
Transient overexpression cell lysate was tested with anti-KIF4A antibody (Mfr. No. H00024137-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)