Learn More
Invitrogen™ KIF4A/KIF4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143922
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human A431 whole cell, human Hela whole cell, human HepG2 whole cell. ICC/IF: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Chromosome-associated kinesin KIF4A is a protein that in humans is encoded by the KIF4A gene. This gene encodes a member of the kinesin 4 subfamily of kinesin related proteins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved in maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
Specifications
KIF4A/KIF4B | |
Polyclonal | |
Unconjugated | |
KIF4B | |
Chromokinesin-A; Chromokinesin-B; KIF4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24137, 285643 | |
-20°C | |
Lyophilized |
ELISA, Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
O95239, Q2VIQ3 | |
KIF4A, KIF4B | |
E.coli-derived human KIF4A/4B recombinant protein (Position: K729-D1174). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.