Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526569
Description
KIF6 Polyclonal Antibody for Western Blot
KIF6 belongs to the kinesin-like protein family. There are four named isoforms. The KIF6 719Arg variant has been associated with an increased risk of coronary disease and an increased response to statin therapy.
Specifications
KIF6 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q6ZMV9 | |
KIF6 | |
KLH conjugated synthetic peptide between 34-62 amino acids from the N-terminal region of human KIF6. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KIF6 | |
C6orf102; D130004B10Rik; D130084M03Rik; dJ1043E3.1; dJ137F1.4; dJ188D3.1; KIF6; kinesin 6; kinesin family member 6; kinesin-like protein KIF6; kinesin-related protein 3; Krp3 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
221458, 319991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction