Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIF7 Monoclonal Antibody (3F8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524382
Description
The antibody detects endogenous KIF7 proteins.
Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
KIF7 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
B7ZNG0, Q2M1P5 | |
KIF7 | |
Recombinant Protein of Human Kif 7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry | |
3F8 | |
Unconjugated | |
KIF7 | |
ACLS; AGBK; EQYK340; HLS2; JBTS12; KIF7; kinesin 7; kinesin family member 7; kinesin-like protein KIF7; UNQ340; UNQ340/PRO539 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
16576, 293047, 374654 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction