Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kilham Rat Virus/KRV-VP1/KRV-VP2 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS4642R
Description
Kilham Rat Virus/KRV-VP1/KRV-VP2 (C-terminus) is involved with the Kilham rat virus (RV), which was found to produce mortality in newborn rats after intracerebral, intravenous, or subcutaneous administration of virus. Both infectious virus and viral hemagglutinins were detected in a variety of tissues and in the blood and urine of experimentally infected rats.Specifications
Kilham Rat Virus/KRV-VP1/KRV-VP2 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
Human |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Liquid | |
KLH conjugated synthetic peptide derived from Ki Kilham Rat Virus/KRV-VP1/KRV-VP2. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title