Learn More
GE Healthcare KiloBase DNA Marker Available on GSA/VA Contract for Federal Government customers only.
Used for quick and precise sizing of double-stranded DNA fragments. GE Healthcare KiloBase DNA Marker consists of 11 easily identified bands when run on agarose gels. Not designed for quantitating DNA content in a sample.
Manufacturer:  GE Healthcare 27400401
Catalog No. 45-002-408
Add to cart
Description
Description
Quick and precise sizing of double-stranded DNA fragments on agarose gels
  • Supplied at 0.5µg per µl in TE buffer (pH 8.0)
  • Sufficient quantity for 100 loadings (using 1µl per loading after mixing with tracking dye)
  • 11 easily-identifiable bands from 0.5 to 10.0kb (when run on a 0.8% agarose gel):
    0.5, 1.0, 1.5, 2.0, 2.5, 3.0, 4.0, 5.0, 6.0, 8.0, and 10.0kb
  • Bands stain with approximately the same intensity, except for the 1.0kb band, which is three times the intensity of the other bands
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
50 μg
0.5 μg/μL
Liquid
DNA Molecular Weight Marker/Ladder